Ethyl cellulose, 300 cps 48% (ethoxyl content)

Supplier: Thermo Scientific Chemicals

Synonyms: Ethylcellulose

23270-0051 23270-5000
TS23270-0051EA 1176 USD
TS23270-0051 TS23270-5000
Ethyl cellulose, 300 cps 48% (ethoxyl content)
Ethyl cellulose
Storage Temperature: Ambient
MDL Number: MFCD00131037
CAS Number: 9004-57-3

Order Now

Specification Test Results

Appearance White Granular powder
Infrared spectrum Conforms
Assay 48 to 49.5 % (ethoxyl)
Viscosity 250 to 350 cP (5 % in 80/20 toluene/ethanol at 25°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR