Emtricitabine

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Amino-5-fluoro-1-((2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl)pyrimidin-2(1H)-one

46207-0010 46207-0050
TS46207-0010EA 126 USD
TS46207-0010 TS46207-0050
Emtricitabine
Emtricitabine
Formula: C₈H₁₀FN₃O₃S
MW: 247.24 g/mol
Melting Pt: 136…140 °C
Storage Temperature: Freezer
MDL Number: MFCD00870151
CAS Number: 143491-57-0

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 153 to 157 °C
Loss on drying ≤0.5 %
HPLC ≥96.0 %
Specific optical rotation -125° to -142° (20 °C, 589 nm) (0.25 % in Methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR