Docetaxel 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Taxotere
45626-0010
45626-2500
TS45626-0010EA
930
USD
TS45626-0010
TS45626-2500
Docetaxel 98%
Docetaxel
Formula:
C₄₃H₅₃NO₁₄ Melting Pt: 186…192 °C Storage Temperature: Ambient |
MDL Number:
MFCD00800737 CAS Number: 114977-28-5 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤6 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...