tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
42797-0010
42797-0050
42797-0250
TS42797-0010EA
41
USD
TS42797-0010
TS42797-0050
TS42797-0250
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate 98%
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate
Formula:
C₁₄H₂₂N₄O₄ Melting Pt: 86…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075122 CAS Number: 152120-54-2 |
Specification Test Results
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 86 to 90 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...