Paclitaxel 99+%

Supplier: Thermo Scientific Chemicals
Danger

32842-0010 32842-0050 32842-0250
TS32842-0010EA 85 USD
TS32842-0010 TS32842-0050 TS32842-0250
Paclitaxel 99+%
Paclitaxel
Formula: C₄₇H₅₁NO₁₄
Melting Pt: 213…216 °C
Storage Temperature: Freezer
MDL Number: MFCD00869953
CAS Number: 33069-62-4

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 203 to 223 °C
HPLC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR