Carbamazepine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5H-Dibenzo[b,f]azepine-5-carboxamide

34436-0250 34436-0010 34436-0050
TS34436-0050EA 94.37 USD
TS34436-0050 TS34436-0250 TS34436-0010
Carbamazepine 98%
Carbamazepine
Formula: C₁₅H₁₂N₂O
MW: 236.27 g/mol
Melting Pt: 189…192 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005073
CAS Number: 298-46-4
EINECS: 206-062-7

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Melting point 189 to 193 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR