Pyridoxal-5'-phosphate monohydrate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Pyridoxal-5-phosphate hydrate, Pyridoxal 5-phosphate monohydrate, Pyridoxal 5'-phosphate monohydrate, (4-Formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate hydrate
22817-0010
22817-0050
22817-0250
TS22817-0010EA
67
USD
TS22817-0010
TS22817-0050
TS22817-0250
Pyridoxal-5'-phosphate monohydrate 99%
Pyridoxal 5'-phosphate monohydrate
Formula:
C₈H₁₀NO₆P·1H₂O Melting Pt: 140…143 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00149414 CAS Number: 41468-25-1 |
Specification Test Results
Appearance | Off-white to yellow Powder |
Infrared spectrum | Conforms |
Water | ≤10 % |
HPLC | ≥98.5 % |
Phosphorus (P) | ≤0.2 % (P inorganic) |
Heavy metals (as Pb) | ≤20 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...