Bacitracin, potency min. 60 Units/mg

Supplier: Thermo Scientific Chemicals
Warning

22610-0050 22610-0250
TS22610-0050EA 163 USD
TS22610-0050 TS22610-0250
Bacitracin, potency min. 60 Units/mg
Bacitracin
Formula: C₆₆H₁₀₃N₁₇O₁₆S₁
MW: 1421.60 g/mol
Melting Pt: 221 °C…225 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00062640
CAS Number: 1405-87-4
EINECS: 215-786-2

Order Now

Specification Test Results

Appearance White to off-white to beige Powder
Infrared spectrum Conforms
Loss on drying ≤5.0 % (1 g, 60°C, 3 h) (P2O5) (not exceeding 0.1 kPa)
Sulfated ash ≤1.0 %
Potency ≥60 IU/mg (on dried substance)
Related substances (Eur. Pharm. (HPLC)) Passes test for composition


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR