Kanamycin sulfate
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Kanamycin Monosulphate
, Kanamycin A
, Kanamycin Monosulfate
45081-0500
45081-0100
45081-1000
TS45081-0500EA
537
USD
TS45081-0500
TS45081-1000
TS45081-0100
Kanamycin sulfate
Kanamycin sulphate
Formula:
C₁₈H₃₈N₄O₁₅S MW: 582.6 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00070253 CAS Number: 25389-94-0 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Assay | ≥760 µg/mg (on dried basis) |
Loss on drying | ≤4.0 % |
Solubility | Clear colorless |
TLC | Complies |
Residue after ignition | ≤1.0 % |
pH | 6.5 to 8.5 (1 % in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...