Ampicillin sodium salt

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt

61177-0050 61177-0250
TS61177-0050EA 101 USD
TS61177-0050 TS61177-0250
Ampicillin sodium salt
Ampicillin sodium salt
Formula: C₁₆H₁₈N₃NaO₄S
MW: 371.39 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00064313
CAS Number: 69-52-3

Order Now

Specification Test Results

Appearance White to tan Powder or crystals
Additional info potency: 845 - 988 mcg/mg
Identification passes test
pH 8.0 to 10.0 (1 % in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR