Glycine-tert-butyl ester hydrochloride(H-Gly-OtBu.HCl) 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Gly-OtBu.HClGlycine tert-butyl ester hydrochlorideH-Gly-OtBu·HCl

29702-0025
TS29702-0025EA 103 USD
TS29702-0025
Glycine-tert-butyl ester hydrochloride(H-Gly-OtBu.HCl) 99%
Glycine tert-butyl ester hydrochloride (H-Gly-OtBu.HCl)
Formula: C₆H₁₄ClNO₂
MW: 167.64 g/mol
Melting Pt: 138…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00058255
CAS Number: 27532-96-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
TLC ≥98.5 %
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR