Chloramphenicol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chloromycetin®(R,R)-ChloramphenicolTega-Cetin

22792-5000 22792-1000 22792-0250
TS22792-0250EA 88.58 USD
TS22792-0250 TS22792-1000 TS22792-5000
Chloramphenicol 98%
Chloramphenicol
Formula: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MW: 323.13 g/mol
Melting Pt: 149…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00078159
CAS Number: 56-75-7
EINECS: 200-287-4
Merck Index: 13,02087

Order Now

Specification Test Results

Appearance White to yellowish-white to greyish-white Crystalline powder
Infrared spectrum Conforms
Melting point 149 to 153 °C
Loss on drying ≤0.5 % (105 °C)
UV spectrophotometry 98.0 to 102.0 % (on dried substance)
Specific optical rotation +18.5° to +20.5° (20 °C, 589 nm) (c=6, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR