Undec-10-enoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

15449-0025 15449-2500 15449-5000
TS15449-0025EA 219 USD
TS15449-0025 TS15449-2500 TS15449-5000
Undec-10-enoic acid 99%
Undec-10-enoic acid
Formula: C₁₁H₂₀O₂
Boiling Pt: 275 °C (760 mmHg)
Melting Pt: 24.5 °C
Density: 0.911 g/cm³
Flash Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD00004442
CAS Number: 112-38-9
EINECS: 203-965-8
Merck Index: 12,09983

Order Now

Specification Test Results

Appearance White to light yellow Low melting solid or liquid
Infrared spectrum Conforms
GC ≥97.0 %
Titration with NaOH 98.5 to 101.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR