Penicillin G sodium salt 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 6-(2-Phenylacetamido)penicillanic acid sodium salt, Benzylpenicillin sodium salt, Sodium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate

45569-0010 45569-0050 45569-0250
TS45569-0010EA 34 USD
TS45569-0010 TS45569-0050 TS45569-0250
Penicillin G sodium salt 98%
Penicillin G sodium salt
Formula: C₁₆H₁₇N₂NaO₄S
Storage Temperature: Ambient
MDL Number: MFCD00069666
CAS Number: 69-57-8

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
pH 5.0 to 7.5
Specific optical rotation +293° to +303° (20°C, 589 nm) (c=2, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR