MES sodium salt 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Sodium 2-Morpholinoethanesulfonate
, Sodium 2-morpholinoethane-1-sulfonate
, MES-Na
, Sodium 3-morpholin-1-ylethylsulfonate
, 2-(N-Morpholino)ethanesulphonic acid sodium salt
, Sodium 3-morpholin-1-ylethylsulphonate
, 2-(N-Morpholino)ethanesulfonic acid sodium salt
, Sodium 2-morpholinoethane-1-sulphonate
, Sodium 2-Morpholinoethanesulphonate
39735-0250
39735-5000
39735-1000
TS39735-5000EA
467
USD
TS39735-5000
TS39735-1000
TS39735-0250
MES sodium salt 99%
MES sodium salt
Formula:
C₆H₁₂NNaO₄S MW: 217.22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00065473 CAS Number: 71119-23-8 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Solubility | (10 % in water) Clear colorless |
UV | (10 % in water) 1 cm cell vs water |
UV | at 260 nm A: ≤0.05 |
UV | at 280 nm A: ≤0.03 |
Titration after Ion exchange | ≥98.5 % |
Heavy metals (as Pb) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...