Nicotinamide 99%

Supplier: Thermo Scientific Chemicals
Warning

12827-0025 12827-1000 12827-5000
TS12827-0025EA 275 USD
TS12827-0025 TS12827-1000 TS12827-5000
Nicotinamide 99%
Nicotinamide
Formula: C₆H₆N₂O
Boiling Pt: 150…160 °C (760 mmHg)
Melting Pt: 128…131 °C
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00006395
CAS Number: 98-92-0
EINECS: 202-713-4
Merck Index: 12,06574

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 128 to 131 °C
Loss on drying ≤0.5 % (4 hrs) (silica gel)
Sulfated ash ≤0.1 %
Readily carbonizable substance Passes test
pH 6.0 to 7.5 (5 % at 20 °C)
Titration with HClO4 ≥98.5 % (on dried substance)
Heavy metals (as Pb) ≤30 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR