myo-Inositol 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Inositolrel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaoli-InositolD-Myo-Inositol

12226-1000 12226-5000 12226-0010 12226-0051
TS12226-1000EA 90 USD
TS12226-1000 TS12226-0010 TS12226-5000 TS12226-0051
myo-Inositol 98+%
myo-Inositol
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 220…228 °C
Storage Temperature: Ambient
MDL Number: MFCD00077932
CAS Number: 87-89-8
EINECS: 201-781-2

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 220 to 228 °C
Loss on drying ≤0.5 %
Solubility (5 % in water) Clear colorless
HPLC ≥98.0 % (ELSD)
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR