Sarcosine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-Sar-OH, N-Methylglycine, Methylaminoacetic acid
15774-5000
15774-0025
15774-1000
TS15774-5000EA
148
USD
TS15774-5000
TS15774-0025
TS15774-1000
Sarcosine 98%
Sarcosine
Formula:
C₃H₇NO₂ Melting Pt: 208 °C (dec.) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004279 CAS Number: 107-97-1 EINECS: 203-538-6 Merck Index: 12,08519 |
Specification Test Results
Appearance | Colorless to white to yellow Powder or crystals |
Infrared spectrum | Conforms |
Water | ≤2 % |
Solubility | (25 % in water) Clear colorless to light yellow |
Titration with HClO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...