Starch Potato Biochemistry, Electrophoresis, hydrolysed

Supplier: Thermo Scientific Chemicals

21512-5000 21512-1000
TS21512-5000EA 197 USD
TS21512-5000 TS21512-1000
Starch Potato Biochemistry, Electrophoresis, hydrolysed
Starch
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤20 %
Gel strength passes test
Residue after ignition ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR