Streptomycin sulfate
Supplier: Thermo Scientific Chemicals
|
Warning
|
45534-0050
45534-0250
45534-1000
TS45534-0050EA
31
USD
TS45534-0050
TS45534-0250
TS45534-1000
Streptomycin sulfate
Streptomycin sulphate
Formula:
C₂₁H₃₉N₇O₁₂·1.5H₂SO₄ MW: 1457.4 g/mol Melting Pt: 250 Storage Temperature: Refrigerator |
MDL Number:
MFCD00037023 CAS Number: 3810-74-0 Merck Index: 12,08983 |
Specification Test Results
Appearance | White to off-white Powder |
Loss on drying | ≤7 % |
Sulfate (SO4) | 18.0 to 21.5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...