L(+)-Arabinose 99% for biochemistry

Supplier: Thermo Scientific Chemicals

Synonyms: Pectin SugarPectinose

36518-0250 36518-1000 36518-5000
TS36518-0250EA 104.47 USD
TS36518-0250 TS36518-1000 TS36518-5000
L(+)-Arabinose 99% for biochemistry
L(+)-Arabinose
Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Melting Pt: 155…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00067709
CAS Number: 5328-37-0
https://us.vwr.com/stibo/web/std.lang.all/24/52/2302452.jpg

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
Solubility (10 % in water) Clear colorless
HPLC ≥98.5 % (ELSD)
UV (1 M in water)
UV at 260 nm A: ≤0.1
UV at 280 nm A: ≤0.1
Residue after ign. (sulfated) ≤0.3 %
Specific optical rotation +102° to +106° (20°C, 589 nm) (10% in water)
Specific optical rotation (after 24 h)
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR