Linoleic acid 60% tech.

Supplier: Thermo Scientific Chemicals
Warning

41319-0025 41319-5000
TS41319-0025EA 317 USD
TS41319-0025 TS41319-5000
Linoleic acid 60% tech.
Linoleic acid
Formula: C₁₈H₃₂O₂
Boiling Pt: 229…230 °C (16 mmHg)
Melting Pt: –5 °C
Density: 0.9 g/cm³
Flash Pt: >112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064241
CAS Number: 60-33-3
EINECS: 200-470-9

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≤1 % C16, 1 double bound
Acid value 195 to 205 mg KOH/g
Iodine value 130 to 148 g I2/100 g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR