L(-)-Tryptophan 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
L-α-Amino-3-indolepropionic acid
, (S)-2-Amino-3-(3-indolyl)propionic acid
, H-Trp-OH
, L-Tryptophan
, Tryptophan
14059-5000
14059-1000
14059-0010
14059-0050
14059-0250
TS14059-0250EA
55.16
USD
TS14059-0250
TS14059-1000
TS14059-0010
TS14059-5000
TS14059-0050
L(-)-Tryptophan 99%
L(-)-Tryptophan
Formula:
C₁₁H₁₂N₂O₂ MW: 204.23 g/mol Melting Pt: 280…285 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064340 CAS Number: 73-22-3 EINECS: 200-795-6 |
Specification Test Results
Appearance | White to yellowish-white Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % (1 g, 105°C) |
Specific optical rotation | -29.0° to -33.0° (20°C, 589 nm) (C=1, H2O) |
Titration with HClO4 | 98.5 to 101.5 % (On dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...