Dithiothreitol (DTT, Cleland's reagent) 99% for biochemistry

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol

16568-0050 16568-0250 16568-0010 16568-1000 16568-5000
TS16568-0050EA 177 USD
TS16568-0050 TS16568-0250 TS16568-0010 TS16568-1000 TS16568-5000
Dithiothreitol (DTT, Cleland's reagent) 99% for biochemistry
Dithiothreitol (DTT, Cleland's reagent)
Formula: C₄H₁₀O₂S₂
Boiling Pt: 125…130 °C (2 mmHg)
Melting Pt: 39…44 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004877
CAS Number: 3483-12-3
EINECS: 222-468-7
UN: 3335
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 38 to 43 °C
Titration Iodimetric ≥98.5 %
UV at 500 nm A: ≤0.035
Appearance of solution (5 % in water) Clear colorless
UV transmittance ≥99 % (c=5, 660 nm, water) 1 cm cell


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR