Dithiothreitol (DTT, Cleland's reagent) 98%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
DL-1,4-Dithiothreitol
, DTT
, cis-2,3-Dihydroxy-1,4-butanedithiol
, (R*,R*)-1,4-Dimercapto-2,3-butandiol
40919-0250
40919-1000
40919-0010
40919-0050
TS40919-0050EA
171.74
USD
TS40919-0050
TS40919-0250
TS40919-1000
TS40919-0010
Dithiothreitol (DTT, Cleland's reagent) 98%, pure
Dithiothreitol (DTT, Cleland's reagent)
Formula:
C₄H₁₀O₂S₂ MW: 154.25 g/mol Boiling Pt: 125…130 °C (2 mmHg) Melting Pt: 39…44 °C Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004877 CAS Number: 3483-12-3 EINECS: 222-468-7 UN: 3335 ADR: 9,III |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Melting point | 38 to 43 °C |
Titration Iodimetric | 97.5 to 102.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...