Dithiothreitol (DTT, Cleland's reagent) 99+% Molecular_Biology DNAse_RNAse_PROTEase_free
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol
32719-0100
32719-0500
32719-0010
TS32719-0100EA
375
USD
TS32719-0100
TS32719-0500
TS32719-0010
Dithiothreitol (DTT, Cleland's reagent) 99+% Molecular_Biology DNAse_RNAse_PROTEase_free
Dithiothreitol (DTT, Cleland's reagent)
Formula:
C₄H₁₀O₂S₂ MW: 154.25 g/mol Boiling Pt: 125…130 °C (2 mmHg) Melting Pt: 39…44 °C Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004877 CAS Number: 3483-12-3 EINECS: 222-468-7 UN: 3335 ADR: 9 III |
|
Specification Test Results
Appearance | White to off-white Solid |
Infrared spectrum | Conforms |
Titration Iodometric | 99.0 to 101.0 % |
UV | (0.1 M in water) |
Dnase,Rnase,Protease activity | none detected |
Heavy metals (as Pb) | ≤5 ppm |
Oxidized DTT(UV) | ≤0.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...