D-(+)-Sucrose ACS

Supplier: Thermo Scientific Chemicals

Synonyms: alpha-D-Glucopyranoside, β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, beta-D-Fructofuranosyl alpha-D-glucopyranoside, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, D-(+)-Saccharose, Sucrose, Sugar

42450-0050 42450-0010 42450-0250
TS42450-0050EA 277 USD
TS42450-0050 TS42450-0010 TS42450-0250
D-(+)-Sucrose ACS
D(+)-Sucrose
Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Melting Pt: 190…192 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystals
Infrared spectrum Conforms
Loss on drying ≤0.03 % (105°C)
Residue after ignition ≤0.01 %
Specific optical rotation +66.3° to +66.8° (25°C, 589 nm) (c=26, H2O)
Chloride (Cl) ≤50 ppm
SO4 and SO3 (as SO4) ≤0.005 %
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Titrable acid ≤0.0008 meq/g
Insoluble matter ≤0.005 %
Invert sugar ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR