D-(+)-Sucrose 99+% for biochemistry DNase-, RNase-, protease-free

Supplier: Thermo Scientific Chemicals

Synonyms: alpha-D-Glucopyranoside, β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, beta-D-Fructofuranosyl alpha-D-glucopyranoside, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, D-(+)-Saccharose, Sucrose, Sugar

41976-0010 41976-0050 41976-0250 41976-2500
TS41976-0010EA 77 USD
TS41976-0010 TS41976-0050 TS41976-0250 TS41976-2500
D-(+)-Sucrose 99+% for biochemistry DNase-, RNase-, protease-free
D(+)-Sucrose
Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Melting Pt: 190…192 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystals
Infrared spectrum Conforms
Loss on drying ≤0.03 %
Solubility (10 % in water) Clear colorless
HPLC ≥99.0 % (ELSD)
Dnase,Rnase,Protease None detected
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR