D-(+)-Sucrose 99.7% for biochemistry
Supplier: Thermo Scientific Chemicals
Synonyms:
alpha-D-Glucopyranoside
, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside
, Sucrose
, D-(+)-Saccharose
, D(+)-Saccharose
, beta-D-Fructofuranosyl alpha-D-glucopyranoside
, β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside
, Sugar
, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru
17714-2500
17714-0010
17714-0250
17714-0050
TS17714-0050EA
124
USD
TS17714-0050
TS17714-0250
TS17714-0010
TS17714-2500
D-(+)-Sucrose 99.7% for biochemistry
D(+)-Sucrose
Formula:
C₁₂H₂₂O₁₁ MW: 342.3 g/mol Melting Pt: 190…192 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006626 CAS Number: 57-50-1 EINECS: 200-334-9 Merck Index: 12,09051 |
Specification Test Results
Appearance | White Coarse crystals |
Infrared spectrum | Conforms |
Loss on drying | ≤0.1 % |
Sulfated ash | ≤0.02 % |
HPLC | ≥99.7 % (ELSD) |
pH | 6.5 to 7.5 (10 % solution) |
Specific optical rotation | (20°C, 589 nm) (c=26, H2O) > +65.9° |
Turbidity | ≤3 NTU (in water) |
Sulfite (as SO2) | ≤15 ppm |
Iron (Fe) | ≤0.5 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.0007 % (in water) |
Reducing sugars | ≤0.04 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...