Agarose, powder, pure for biochemistry (Mr = - 0.17)
Supplier: Thermo Scientific Chemicals
23902-5000
23902-1000
23902-0250
TS23902-5000EA
986
USD
TS23902-5000
TS23902-0250
TS23902-1000
Agarose, powder, pure for biochemistry (Mr = - 0.17)
Agarose
Storage Temperature:
Ambient |
MDL Number:
MFCD00081294 CAS Number: 9012-36-6 EINECS: 232-731-8 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤7 % (5 g, 110°C, 24 h) |
Gel strength | ≥2000 g/cm² (1.5 % gel) |
Gel point | 34.5 to 37.5°C (1.5 % gel) |
Electroendosmosis | 0.16 to 0.19 (-Mr) |
Ash | ≤0.5 % |
Sulfate (SO4) | ≤0.25 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...