Adenine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 7H-Purin-6-amine6-AminopurinePurin-6-amine

14744-0250 14744-5000 14744-1000
TS14744-1000EA 332 USD
TS14744-1000 TS14744-0250 TS14744-5000
Adenine 99%
Adenine
Formula: C₅H₅N₅
MW: 135.13 g/mol
Melting Pt: 360…365 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00041790
CAS Number: 73-24-5
EINECS: 200-796-1
UN: 2811
ADR: 6.1,III
Merck Index: 13,00152

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 5 hrs)
Sulfated ash ≤0.1 %
HPLC ≥98.5 %
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR