D-(-)-Arabinose 99+%
Supplier: Thermo Scientific Chemicals
Synonyms:
D-(-)-Arabinose, (2S,3R,4R)-2,3,4,5-Tetrahydroxypentanal
16145-0250
16145-1000
TS16145-0250EA
62
USD
TS16145-0250
TS16145-1000
D-(-)-Arabinose 99+%
D(-)-Arabinose
Formula:
C₅H₁₀O₅ MW: 150.13 g/mol Melting Pt: 160 Storage Temperature: Ambient |
MDL Number:
MFCD00135608 CAS Number: 10323-20-3 EINECS: 233-708-5 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Water | ≤0.5 % (K.F.) |
Sulfated ash | ≤0.1 % |
HPLC | ≥99.0 % (ELSD) |
Specific optical rotation | -101.0° to 106.0° (20°C, 589 nm) (c=4, H2O) (after 24 h) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...