HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid 99+% for molecular biology DNase-, RNase-, protease-free

Supplier: Thermo Scientific Chemicals

Synonyms: HEPES free acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulfonic acid, 4-(2-Hydroxyethyl)piperazin-1-ylethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid), 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid

32985-0500 32985-5000
TS32985-0500EA 195 USD
TS32985-0500 TS32985-5000
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid 99+% for molecular biology DNase-, RNase-, protease-free
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)
Formula: C₈H₁₈N₂O₄S
MW: 238.31 g/mol
Melting Pt: 209…215 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006158
CAS Number: 7365-45-9
EINECS: 230-907-9
Merck Index: 13,04672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C)
UV (0.5 M in water)
pH 5 to 6.5 (0.5 M in water)
Titration with NaOH ≥99.0 % (On dry substance)
Dnase,Rnase,Protease activity none detected
Sulfate (SO4) ≤500 ppm
Iron (Fe) ≤5 ppm
Potassium (K) ≤1000 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR