Bisacrylamide 99+% for biochemistry, for electrophoresis

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N,N’-MethylenebisacrylamideN,N'-MethylenebisacrylamideBis (N,N'-methylenebisacrylamide)N,N-MethylenebisacrylamideN,N'-MethylenediacrylamideBIS

16479-5000 16479-1000 16479-0250
TS16479-1000EA 131 USD
TS16479-1000 TS16479-5000 TS16479-0250
Bisacrylamide 99+% for biochemistry, for electrophoresis
Bisacrylamide
Formula: (H₂C=CHCONH)₂CH₂
MW: 154.17 g/mol
Melting Pt: 185 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00008625
CAS Number: 110-26-9
EINECS: 203-750-9
https://us.vwr.com/stibo/web/std.lang.all/70/49/2297049.jpg

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals or needles
Infrared spectrum Conforms
HPLC ≥99.0 %
UV (1 % in water) 1 cm cell
UV at 290 nm A: ≤0.25
pH 5 to 7 (1 % in water)
Acrylic acid ≤0.05 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR