Bisacrylamide 96%, Extra Pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N,N’-MethylenebisacrylamideN,N'-MethylenebisacrylamideBis (N,N'-methylenebisacrylamide)N,N-MethylenebisacrylamideN,N'-MethylenediacrylamideBIS

16380-5000 16380-1000 16380-0025
TS16380-5000EA 116 USD
TS16380-5000 TS16380-1000 TS16380-0025
Bisacrylamide 96%, Extra Pure
Bisacrylamide
Formula: (H₂C=CHCONH)₂CH₂
MW: 154.17 g/mol
Melting Pt: 185 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00008625
CAS Number: 110-26-9
EINECS: 203-750-9
https://us.vwr.com/stibo/web/std.lang.all/70/49/2297049.jpg

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
HPLC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR