Urea 99.5% for molecular biology DNase-, RNase-, protease-free
Supplier: Thermo Scientific Chemicals
32738-0010
32738-0050
TS32738-0010EA
130
USD
TS32738-0010
TS32738-0050
Urea 99.5% for molecular biology DNase-, RNase-, protease-free
Urea
Formula:
NH₂CONH₂ Melting Pt: 131…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008022 CAS Number: 57-13-6 EINECS: 200-315-5 REACH: 01-2119463277-33 Merck Index: 13,09935 |
|
Specification Test Results
Appearance | White Solid |
Assay | ≥99.5 % |
UV | (8 M in water) |
Dnase,Rnase,Protease activity | none detected |
Chloride (Cl) | ≤5 ppm |
Iron (Fe) | ≤10 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...