Sodium L(+)-ascorbate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Sodium ascorbate
, L(+)-Ascorbic acid sodium salt
, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate
, L-Ascorbic acid sodium salt
, Acidum ascorbicum sodium salt
35268-1000
35268-0025
35268-5000
35268-0050
TS35268-1000EA
83.4
USD
TS35268-1000
TS35268-0025
TS35268-0050
TS35268-5000
Sodium L(+)-ascorbate 99%
Sodium L(+)-ascorbate
Formula:
C₆H₇NaO₆ MW: 198.11 g/mol Melting Pt: 219…221 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00082340 CAS Number: 134-03-2 EINECS: 205-126-1 |
Specification Test Results
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Titration Iodimetric | 98.5 to 101.5 % |
Solubility | (5% in water) Clear colorless to light yellow |
Specific optical rotation | +103° to +108° (20°C, 589 nm) (c=5, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...