Sodium chloride 99.85% Molecular_Biology DNAse_RNAse_PROTEase_free

Supplier: Thermo Scientific Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

32730-0010 32730-0025
TS32730-0010EA 100 USD
TS32730-0010 TS32730-0025
Sodium chloride 99.85% Molecular_Biology DNAse_RNAse_PROTEase_free
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Appearance White Crystalline solid
Titration Argentometric ≥99.80 %
UV at 280 nm A: ≤0.01
Dnase,Rnase,Protease activity none detected
Iodide (I) ≤20 ppm
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤4 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤2 ppm
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤50 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR