Lithium chloride 99% for molecular biology

Supplier: Thermo Scientific Chemicals
Warning

44904-5000 44904-1000
TS44904-5000EA 240 USD
TS44904-5000 TS44904-1000
Lithium chloride 99% for molecular biology
Lithium chloride
Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1382
Melting Pt: 614
Density: 2.06 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
Merck Index: 13,05550

Order Now

Specification Test Results

Appearance White Powder
Titration Argentometric ≥98.5 %
Dnase,Rnase,Protease activity Not detected
Iron (Fe) ≤0.0005 %
Magnesium (Mg) ≤0.005 %
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR