Sodium dodecyl sulfate (SDS) 99+% for molecular biology DNase-, RNase-, protease-free

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium dodecyl sulfateLauryl sulfate sodium saltSodium lauryl sulfateDodecyl sulfate sodium saltDodecyl sulphuric acid sodium saltLauryl sulphate sodium saltSodium lauryl sulphateDodecyl sulphate sodium saltDodecyl sulfuric acid sodium salt

32731-1000 32731-5000 32731-0025
TS32731-0025EA 1520 USD
TS32731-0025 TS32731-1000 TS32731-5000
Sodium dodecyl sulfate (SDS) 99+% for molecular biology DNase-, RNase-, protease-free
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Melting Pt: 206 °C
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,III
Merck Index: 14,08636
https://us.vwr.com/stibo/web/std.lang.all/54/91/735491.jpg

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Assay ≥99.0 %
UV (0.1 M in water)
UV at 260 nm A: ≤0.05
UV at 280 nm A: ≤0.05
Dnase,Rnase,Protease activity None detected
Chloride (Cl) ≤1000 ppm
Phosphate (PO4) ≤5 ppm
Copper (Cu) ≤5 ppm
Lead (Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR