Arachidic acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Eicosanylic acid, Arachidinic acid, Icosanoic acid, Eicosanoic acid

11772-0010 11772-0100 11772-0250
TS11772-0010EA 34 USD
TS11772-0010 TS11772-0100 TS11772-0250
Arachidic acid 98%
Arachidic acid
Formula: C₂₀H₄₀O₂
MW: 312.54 g/mol
Boiling Pt: 328
Melting Pt: 75.5
Storage Temperature: Ambient
MDL Number: MFCD00002755
CAS Number: 506-30-9
EINECS: 208-031-3
Merck Index: 12,00802

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 73 to 78 °C
GC ≥97.5 %
Titration with TBAH 97.5 to 102.5 %
Solubility Passes test
Sensitivity (to sulfanilamide) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR