Ninhydrin

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 2,2-Dihydroxyindan-1,3-dionTrioxohydrindene monohydrateNinhydrin monohydrate2,2-Dihydroxy-1,3-indanedioneIndanetrione hydrate2,2-Dihydroxyindane-1,3-dione1,2,3-Indantrione monohydrate

0210245090
76461-442EA 1246.01 USD
76461-442
Ninhydrin
Ninhydrin
Formula: C₉H₆O₄
MW: 178.14 g/mol
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 250…258 °C
Density: 0.862 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003791
CAS Number: 485-47-2
EINECS: 207-618-1
Merck Index: 13,06582

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR