Crystal violet

Supplier: bioMerieux
Danger

Synonyms: Basic Violet 3GentiavermMethylrosanilide chlorideHexamethyl VioletVianinGentian violetMethyl violet 10BCI 42555Hexamethyl-p-rosaniline chlorideN-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanaminium chloride4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chlorideDimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-yliden)ammoniumchloridViocid

55545
76459-552EA 79.57 USD
76459-552
Crystal violet
Crystal Violet
Formula: C₂₅H₃₀ClN₃
MW: 407.99 g/mol
Melting Pt: 189…194 °C
Density: 1.19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011750
CAS Number: 548-62-9
EINECS: 208-953-6
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR