Dr. FuelCell® Science Kit
Supplier: KLINGER EDUCATION PROD CORP SE
355 350
470335-980KT 693.89 USD
470335-980 470335-986
Dr. FuelCell® Science Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Dr FuelCell® Science Kit is an extensive experiment set for the subject of renewable energies. Flexible components, 20 pre-configured experiments and detailed supplementary material make it a complete package for teaching physics and chemistry in grades 9 to 12.
  • Curriculum oriented instruction material (Grades 9 to 12)
  • 20 pre-configured experiments for individual or group work
  • Immediately ready to use, no additional materials needed
  • Robust and user-friendly components
  • Practical and easy-to-operate load measurement box for measuring current and voltage
  • Expandable, for gaining expert knowledge of fuel cells
The Dr. Fuel Cell® Science Kit is an extensive experiment set for the subject of renewable energies. Flexible components, 20 pre-configured experiments and detailed supplementary material make it a complete solution for teaching physics and chemistry in grades 9 to 12. The Dr. FuelCell® Science Kit reproduces a complete solar hydrogen energy cycle. It makes it possible to approach the subject of renewable energies both as a complete cycle and at the level of the single technologies of photovoltaics and fuel cell.
The curriculum-oriented documentation of the Dr. FuelCell® Science Kit contains more than 20 preconfigured experiments with technical background information as an ideal preparation aid for instruction. The system is suitable for content from the curricula of physics and chemistry for the lower and upper secondary level. Topics include: Molecules and chemical reactions; Reaction speeds; Thermodynamics; Electrochemistry; Energy conversion and efficiency; Measuring and interpreting characteristic curves; Planning and implementation of scientific experiments.
Ordering information: The package includes a solar panel, an electrolyzer with two graduated storage cylinders, a fuel cell and a load measurement box. A methanol fuel cell and a take-apart fuel cell are included in the Complete science kit.