1-Naphtholbenzeine in acetic acid indicator

Supplier: Thermo Scientific Chemicals

Synonyms: alpha-Naphtholbenzeinα-Naphtholbenzein4-(α-(4-Hydroxy-1-naphthyl)benzylidene)naphthalen-1(4H)-one4-Hydroxy-α-(4-hydroxynaphthyl)-α-phenylnaphthalene-1-methanolα-Naphtholbenzeinep-Naphtholbenzeine1-NaphtholbenzeinBis(4-hydroxy-1-naphthyl)phenylmethanolp-Naphtholbenzein

61131-5000
TS61131-5000EA 262 USD
TS61131-5000
1-Naphtholbenzeine in acetic acid indicator
1-Naphtholbenzeine
Formula: C₂₇H₁₈O₂
MW: 374.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078492
CAS Number: 145-50-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance clear brownish-red liquid
Sensitivity passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR