Diethyl carbonate, anhydrous 99%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Warning

43613-0010 43613-1000
TS43613-0010EA 304 USD
TS43613-0010 TS43613-1000
Diethyl carbonate, anhydrous 99%, AcroSeal™
Diethyl carbonate
Formula: C₅H₁₀O₃
Boiling Pt: 126…128 °C (760 mmHg)
Melting Pt: –43 °C
Density: 0.975 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00009107
CAS Number: 105-58-8
UN: 2366
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3830 to 1.3850 (20°C, 589 nm)
Water ≤0.1 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR