Sulfaguanidine 98%

Supplier: Thermo Scientific Chemicals
Warning

43130-0250 43130-1000 43130-5000
TS43130-0250EA 34 USD
TS43130-0250 TS43130-1000 TS43130-5000
Sulfaguanidine 98%
Sulfaguanidine
Formula: C₇H₁₀N₄O₂S
Melting Pt: 190.5…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00038136
CAS Number: 57-67-0

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 189 to 193 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR