Triisopropyl borate 98+%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

42714-1000 42714-8000
TS42714-1000EA 60 USD
TS42714-1000 TS42714-8000
Triisopropyl borate 98+%, AcroSeal™
Triisopropyl borate
Formula: C₉H₂₁BO₃
Boiling Pt: 139 °C (1013 hPa)
Melting Pt: –59 °C
Density: 0.814 g/cm³
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00008872
CAS Number: 5419-55-6
UN: 2616
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3755 to 1.3775 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR