Rose Bengal 85%, pure, certified

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium 4,5,6,7-tetrachloro-2',4',5',7'-tetraiodo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Food Red No. 105, CI 45440, Acid red 94, Aniline Rose, Rose Bengal sodium

41915-0250 41915-1000
TS41915-0250EA 105 USD
TS41915-0250 TS41915-1000
Rose Bengal 85%, pure, certified
Rose Bengal
Formula: C₂₀H₂Cl₄I₄O₅Na₂
Storage Temperature: Ambient
MDL Number: MFCD00151169
CAS Number: 632-69-9

Order Now

Specification Test Results

Appearance Red-brown Powder
Infrared spectrum Conforms
Dye content ≥80 %
UV Lambda max 544 to 550 nm
Absorption Ratio P-15/P+15 1.21 to 1.95
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR