Potassium hydrogen phthalate 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: Potassium phthalate monobasic, Potassium biphthalate, Potassium Acid Phthalate, KHP, Phthalic Acid Monopotassium Salt, Phthalic acid potassium salt

41795-0025 41795-5000
TS41795-0025EA 294 USD
TS41795-0025 TS41795-5000
Potassium hydrogen phthalate 99+%
Potassium hydrogen phthalate
Formula: HOOCC₆H₄COOK
MW: 204.22 g/mol
Melting Pt: 295…300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013070
CAS Number: 877-24-7
EINECS: 212-889-4
Merck Index: 13,07694
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4652479e822d1591ac7eb5e29f892b8c94588ad4&vid=75071da7f30b07f7bd077699a3b54ec89d80f0b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Titration with NaOH ≥99.0 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR